Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazinamide, 98%

Catalog No. AAJ6682214
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6682214 25 g
AAJ6682209 10 g
AAJ6682222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6682214 Supplier Thermo Scientific Chemicals Supplier No. J6682214
Add to Cart

CAS: 98-96-4 | C5H5N3O | 123.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.115
MDL Number MFCD00006132
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N
PubChem CID 1046
ChEBI CHEBI:45285
IUPAC Name pyrazine-2-carboxamide
SMILES C1=CN=C(C=N1)C(=O)N

Specifications

Type Pyrazinamide
Physical Form Powder
Melting Point 189°C to 191°C
Packaging Plastic Bottle
Quantity 25 g
Flash Point >110°C (230°F)
Beilstein 112306
Synonym Pyrazinoic acid amide,Pyrazinecarboxamide
Solubility Information Soluble in water /n
Formula Weight 123.11
Percent Purity 98%
Chemical Name or Material Pyrazinamide
Show More Show Less
EINECSNumber 202-717-6
RTECSNumber UQ2275000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.