Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazole-3,5-dicarboxylic Acid Monohydrate 98.0+%, TCI America™
SDP

Catalog No. P104825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P104825G 25 g
P10485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P104825G Supplier TCI America Supplier No. P104825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3112-31-0
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.097
MDL Number MFCD00005235
InChI Key YDMVPJZBYSWOOP-UHFFFAOYSA-N
Synonym 3,5-pyrazoledicarboxylic acid, pyrazole-3,5-dicarboxylic acid, pyrazole-3,5-dicarboxylic acid monohydrate, 3,5-pyrazol dicarboxylic acid, 3,5-pyrazole dicarboxylic acid, zlchem 726, 3,5-dicarboxypyrazole, acmc-20a0hc, ksc223c6j, 3,5-pyrazole-dicarboxylic acid
PubChem CID 76559
IUPAC Name 1H-pyrazole-3,5-dicarboxylic acid
SMILES C1=C(NN=C1C(=O)O)C(=O)O

Specifications

Melting Point 295°C
Color White
Quantity 25 g
Formula Weight 156.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazole-3,5-dicarboxylic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.