Learn More
Pyrethroids in Ground Water, WS Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017329 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7329 2 mL
1 options
Catalog No. 11-101-7329 Supplier MilliporeSigma Supelco Supplier No. PE13172ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL
Concentration or Composition (by Analyte or Components) cis-Permethrin 0.5-120 ng/L; Cypermethrin (sum of 4 isomers) 0.5-500 ng/L; Deltamethrin 0.5-120 ng/L; Flumethrin (sum of 2 isomers) 0.5-300 ng/L; Pentachlorophenol 0.5-120 ng/L; trans Permethrin 0.5-120 ng/L