Learn More
Cole-Parmer Pyrex 51395-500 Brand 51395 UV-Blocking Low Actinic Media Bottle, 500 mL, 4/cs
Supplier:  Cole-Parmer 3457423
Encompass_Preferred
Made from low actinic, code no. 7740 borosilicate glass with red stain, these heavy-duty bottles are ideal for storing/transporting chemicals sensitive to light in the 3000 to 5000 Å range (300 to 500 nm).
Bottles include a linerless, one-piece autoclavable, orange polypropylene, plug-seal screw cap with drip-free pouring ring. Glass bead indicates full capacity line. Bottles feature white enamel graduations and marking spots. Replacement caps and pouring rings are available (sold separately).
Catalog No. NC9863147