Learn More
Corning™ PYREX™ Cylindrical Separatory Funnel, Graduated, PTFE Standard Stopcock, Glass Standard Taper Stopper
Microfinish PTFE stopcock plugs reduce freezing problems and eliminate contaminating greases
$2330.00
Specifications
Material | Borosilicate Glass |
---|---|
Graduated | Yes |
Includes | Standard stopcock and stopper |
Joint Number | 24/40 |
Product Type | Cylindrical Separatory Funnel |
Description
This funnel is graduated to show approximate capacity, has a micro-finish PTFE stopcock, and the strong, lightweight PYREX stopper.
For stopper only, see Cat. No. 7650
Order Info
Approximate stem length, 65mm. Replacement stoppers are available; see Stoppers.
Specifications
Borosilicate Glass | |
Standard stopcock and stopper | |
Cylindrical Separatory Funnel |
Yes | |
24/40 |