Learn More
Corning™ PYREX™ Fritted Disc Vertical Gas Dispersion Tubes
Catalog No. 22124487 Shop All Corning Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
22-124487 Borosilicate Glass
1 options
Catalog No. 22-124487 Supplier Corning™ Supplier No. 3953030C
Used to disperse or scrub gas in liquids
  • 30mm diameter tubes
  • Coarse porosity (40 to 60μm) fritted disc at angle of approximately 15°
  • Permits small gas bubbles to rise from disc surface without excessive formation of large bubbles
  • Performance comparable to that of horizontal disc (No. 39525)
  • Can be used in test tubes or flasks since large opening is not required
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass