Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Rimless Culture Tubes with Flat Bottoms
SCI_ED

Catalog No. S67133 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
47 mL
Length (Metric):
120 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Length (Metric)
S67133 47 mL 120 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67133 Supplier Corning™ Supplier No. 985025X/EMD

Ideal for plant tissue culture and general culture work

Ideal for plant tissue culture and general culture work. Available in packs of 36 except S67135 which is available in packs of 24.
•    Walls and bottoms are uniform thickness
•    Rimless top for greater ease in plugging and rack storage
•    Flat bottom facilitates use with agar slants

Two-position polypropylene closures available separately. Available in cases of 100.
•    Autoclavable
•    Position closure OPEN for gas exchange or CLOSED for humidified environment
•    Internal drip ring minimizes contamination

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 47 mL
Length (Metric) 120 mm
Material Borosilicate Glass
Disposable Reusable
For Use With For plant tissue culture and general culture work
Label Type None
Rimmed No
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Diameter (Metric) 25 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.