Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ VISTA™ Class A Cylinder
SCI_ED

Catalog No. S76102E Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S76102E 250 mL
S76102C 50 mL
S76102A 10 mL
S76102B 25 mL
S76102D 100 mL
S76102F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S76102E Supplier Corning™ Supplier No. 70024250/EMD
This item is not returnable. View return policy

Calibrated To Deliver

  • Blue enamel graduations
  • Capacity is in accordance with ASTM E-1272
  • 10mL size has a funnel top for easier filling

TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass
Capacity (Metric) 250 mL
Scale Single
Sterility Non-sterile
Calibration To Deliver
Scale Color Blue
Class Class A
Certifications/Compliance ASTM E1272
Product Type Graduated Cylinder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.