Description
- Provide precise volume measurement
- Graduation line is sharp and permanent
- Large blue block letters make the inscription easy to read
Compliance
Manufactured to Class A tolerances as established by ASTM™ E-694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.
Specifications
Specifications
| Closure Material | Glass |
| Capacity (Metric) | 10 mL |
| Material | Glass, Borosilicate Glass |
| Neck Style | Long |
| Graduated | Yes |
| Bottom Shape | Flat |