Learn More
Apogee Instruments Inc PYRGEOMETER UPWARD-LOOKING
SDP
Supplier:  Apogee Instruments Inc SL510SS
Encompass_Preferred
Pyrgeometer Upward-LookingThe SL-510 is an upward-looking thermopile pyrgeometer. The sensor incorporates a filter, blackbody thermopile detector, and thermistor with a rugged, self-cleaning sensor housing design
Catalog No. NC2292096
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.