Learn More
AdipoGen Pyribenzoxim
SDP
Supplier:  AdipoGen CDXP0132M100
Encompass_Preferred
Chemical. CAS 168088-61-7. Formula C32H27N5O8. MW 609.59. Synthetic. Unclassified herbicide. Broad-spectrum post-emergence herbicide used to control grasses and polygonums in rice, wheat and other crops. Its mode of action is via the inhibition of acetolactate synthase ALS acetohydroxyacid synthase AHAS. Compound can be used as analytical reference material.
Catalog No. 50-256-0231
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.