Learn More
AdipoGen Pyributicarb
SDP
Supplier:  AdipoGen CDXP0129M100
Encompass_Preferred
Chemical. CAS 88678-67-5. Formula C18H22N2O2S. MW 330.4. Synthetic. Thiocarbamate herbicide with fungicidal activity. Inhibits 4-hydroxyphenyl-pyruvatedioxygenase 4-HPPD in plants that causes bleaching. And targets squalene-epoxidase in sterol biosynthesis in fungus. Compound can be used as analytical reference material.
Catalog No. 50-256-0235
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.