Learn More
AdipoGen Pyridafol
SDP
Supplier:  AdipoGen CDXP0133M100
Encompass_Preferred
Chemical. CAS 40020-01-7. Formula C10H7ClN2O. MW 206.63. Synthetic. Pyridazine herbicide. Mode of action is through the inhibition of photosynthetic electron transport at the photosystem II. Used against broad-leaved weeds including nightshade, cleavers, nettles, chickweed and grasses including barnyard grass, crabgrass, foxtail. Compound can be used as analytical reference material.
Catalog No. 50-256-0228
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.