Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridazine 99.0+%, TCI America™
SDP

Catalog No. P061625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P061625G 25 g
P06165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P061625G Supplier TCI America Supplier No. P061625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 289-80-5
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006463
InChI Key PBMFSQRYOILNGV-UHFFFAOYSA-N
Synonym orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
PubChem CID 9259
ChEBI CHEBI:30954
IUPAC Name pyridazine
SMILES C1=CC=NN=C1

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 208°C
Quantity 25 g
Formula Weight 80.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyridazine
RTECSNumber UR5267000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.