Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, 99.0% min., ACS, MilliporeSigma™

Catalog No. MPX20205 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX20205 4 L
MPX202011 100 mL
MPX20206 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MPX20205 Supplier MilliporeSigma Supplier No. PX20205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Color Colorless
Density 0.982g/cm3 (20°C)
Quantity 4 L
Explosion Limit 1.7 to 10.6%(V)
Ignition Point 482°C
Physical Form Liquid
Assay Percent Range 99.0% min.
Vapor Pressure 20 hPa (20°C)
Formula Weight 79.1
Chemical Name or Material Pyridine, SeccoSolv™, Dried
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.