Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, 99+%, for analysis

Catalog No. AC290860100
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290860100 10 L Plastic drum
AC290861000 100 mL Glass bottle
AC290860010 1 L Glass bottle
AC290860025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC290860100 Supplier Thermo Scientific Chemicals Supplier No. 290860100
Add to Cart

CAS: 110-86-1 | C5H5N | 79.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.1
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42.0°C
Density 0.9780g/mL
Boiling Point 115.0°C to 116.0°C
Flash Point 17°C
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Refractive Index 1.5085 to 1.5105
Quantity 10 L
Beilstein 20,181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Specific Gravity 0.978
Merck Index 15,8081
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Viscosity 0.95 mPa.s (20°C)
Formula Weight 79.1
Percent Purity 99+%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Pyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 203-809-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.