Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, ACS Reagent, Reagents

Catalog No. 01337777
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Glass Bottle
Safety Coated Glass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-777 4 L Safety Coated Glass
01-337-781 100 mL Glass Bottle
01-337-780 1 L Safety Coated Glass
01-337-778 4 L Glass Bottle
01-337-776 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-777 Supplier Reagents Holdings Llc Supplier No. C12940004C1
Add to Cart
Add to Cart

Pyridine, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.10
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym Azabenzene, Azine
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point <-29°C
Color Colorless
Boiling Point 115°C
Packaging Safety Coated Glass
Quantity 4 L
Specific Gravity 0.982
Grade ACS
Physical Form Liquid
Chemical Name or Material Pyridine, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.