Learn More
Pyridine-Barbituric Acid Reagent, For Cyanide, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Pyridine-Barbituric Acid Reagent |
| Color | Amber |
| Name Note | For Cyanide |
| CAS Max % | 62, 30, 5.6, 2.4 |
| CAS | 110-86-1, 67-52-7, 7647-01-0 |
| Linear Formula | C5H5N |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Solubility Information | Soluble in water |
| Formula Weight | 79.1 |
| Grade | Certified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.