Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine,Solstice

Catalog No. 60048571 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-571 4 x 4 L Glass Bottle
60-090-481 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-571 Supplier Solstice Supplier No. BB3334
Add to Cart
Add to Cart
Request a Quote

BioSyn™, for biosynthesis and applications requiring low water content, >99.8%

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.10
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Quantity 4 x 4 L
Flash Point 20 °C
Packaging Glass Bottle
Physical Form Liquid, Clear
Refractive Index 1.5085 - 1.5109
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.