Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine - Sulfur Trioxide Complex 95.0+%, TCI America™
SDP

Catalog No. P0998100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0998100G 100 g
P0998500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0998100G Supplier TCI America Supplier No. P0998100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26412-87-3
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00012437
InChI Key UDYFLDICVHJSOY-UHFFFAOYSA-N
Synonym pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
PubChem CID 168533
IUPAC Name pyridine; sulfonylideneoxidane
SMILES O=S(=O)=O.C1=CC=NC=C1

Specifications

Color White-Yellow
Quantity 100 g
UN Number 3261
Formula Weight 159.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine - Sulfur Trioxide Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.