Learn More
Apexbio Technology LLC Pyridone 6 457081-03-7 5mg
SDP
Supplier:  Apexbio Technology LLC A37415
Encompass_Preferred
Pyridone 6 (CAS 457081-03-7) is a reversible ATP-competitive inhibitor of Janus kinases (JAKs) exhibiting high selectivity with IC50 values of 15 nM for JAK1 1 nM for JAK2 5 nM for JAK3 and 1 nM for TYK2 It demonstrates markedly reduced activity against other kinases (IC50 130 nM) Pyridone 6 disrupts JAK-STAT signaling suppressing cytokine-induced STAT3 phosphorylation and reducing cellular proliferation in relevant models such as B9 and U266 cells Its utility extends to animal models where it has been employed to study immune modulation and inflammation including atopic dermatitis by inhibiting Th cell differentiation and mast cell function
Catalog No. 50-224-0167
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.