Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridostigmine bromide, 98%

Catalog No. AC461190050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461190050 5 g
AC461190010 1 g
AC461190250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC461190050 Supplier Thermo Scientific Chemicals Supplier No. 461190050
Add to Cart

CAS: 101-26-8 | C9H13BrN2O2 | 261.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-26-8
Molecular Formula C9H13BrN2O2
Molecular Weight (g/mol) 261.12
MDL Number MFCD00079283
InChI Key VNYBTNPBYXSMOO-UHFFFAOYSA-M
Synonym pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide
PubChem CID 7550
IUPAC Name (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1

Specifications

Melting Point 154.0°C
Loss on Drying 2% max. (1 g, 105°C)
Infrared Spectrum Authentic
Quantity 5 g
Merck Index 15,809
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, chloroform, insoluble in ether, benzene, acetone
Formula Weight 261.12
Percent Purity 98%
Chemical Name or Material Pyridostigmine bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause an allergic skin reaction.
Precautionary Statement GHS P Statement Rinse mouth. Wear protective gloves. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 202-929-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.