Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridostigmine Bromide 98.0+%, TCI America™
SDP

Catalog No. P133925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P133925G 25 g
P13395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P133925G missing translation for 'mfr' TCI America missing translation for 'supplierNo' P133925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 101-26-8
Molecular Formula C9H13BrN2O2
Molecular Weight (g/mol) 261.12
MDL Number MFCD00079283
InChI Key VNYBTNPBYXSMOO-UHFFFAOYSA-M
Synonym pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide
PubChem CID 7550
IUPAC Name 3-[(dimethylcarbamoyl)oxy]-1-methylpyridin-1-ium bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1

Specifications

Melting Point 154°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 261.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridostigmine Bromide
EINECSNumber (9)-0648
RTECSNumber UU5270000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.