Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine Hydrochloride (B6), MilliporeSigma™ Supelco™

Catalog No. 111014758 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4758 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4758 Supplier MilliporeSigma Supelco Supplier No. 47862
Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 58-56-0
Physical Form Neat
Linear Formula C8H11 NO3 · HCl
MDL Number MFCD00012807
Melting Point 214°C to 215°C
Synonym Pyridoxine hydrochloride; Adermine hydrochloride; PN HCl; Pyridoxol hydrochloride; Vitamin B6 hydrochloride
Type Analytical Standard
Quantity 1000 mg
Formula Weight 205.64
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.