Learn More
Lakeland Industries Pyrolon™ Plus 2 Disposable Flame Resistant Lab Coats
Catalog No. 19813328
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-328 5X-Large
19-813-330 Medium
19-813-329 Large
19-813-332 X-Large
19-813-325 2X-Large
19-813-326 3X-Large
19-813-327 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-813-328 Supplier Lakeland Industries Supplier No. 7101B5X
Add to Cart
Add to Cart
Disposable flame resistant/ARC lab coat also protects from dirt, grease, oil, and other harmful contaminants.
  • Disposable flame resistant labcoats protect primary flame resistant garments from dirt, grease, and light aerosol spray.
    • Disposable secondary flame resistant protection
    • Designed to be worn over primary flame resistant clothing
    • Oil and water replelant
  • Disposable flame resistant lab coat designed to keep primary flame resistant garments free of dirt, grease, oil, and other contaminants. Lab coat features long sleeves, 2 pockets, and snap closure.
  • Certifications: Meets NFPA 2113 requirements for section 5.1.9.
Specifications
Product Type Flame-resistant Lab Coat
Size 5X-Large
Gender Unisex
Material Wood Pulp Spunlace, FR-treated Polyethylene
Color Blue
Closure Type Snaps
Disposable Single-use
Sterility Non-sterile
Resistance Flame
No. of Pockets 2
Model 7101B
Certifications/Compliance ANSI/ISEA 203 Certified, NFPA 2113