Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyromellitic Diimide 97.0+%, TCI America™
SDP

Catalog No. P115325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P115325G 25 g
P11535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P115325G Supplier TCI America Supplier No. P115325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2550-73-4
Molecular Formula C10H4N2O4
Molecular Weight (g/mol) 216.152
MDL Number MFCD00005004
InChI Key UGQZLDXDWSPAOM-UHFFFAOYSA-N
Synonym pyromellitic diimide, pyromellitimide, pyromellitic acid diimide, benzo 1,2-c:4,5-c' dipyrrole-1,3,5,7 2h,6h-tetrone, unii-32xa0w314g, 1,2,4,5-benzenetetracarboxylic acid diimide, 1,2,4,5-benzenetetracarboxylic 1,2:4,5-diimide, benzene-1,2,4,5-tetracarboxylic diimide, pyrrolo 3,4-f isoindole-1,3,5,7-tetrone, pyrrolo 3,4-f isoindole-1,3,5,7 2h,6h-tetrone
PubChem CID 75696
IUPAC Name pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
SMILES C1=C2C(=CC3=C1C(=O)NC3=O)C(=O)NC2=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 216.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyromellitic Diimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.