Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyronin B, 50% dye content, Thermo Scientific™

Catalog No. AC191390050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC191390050 5 g
AC191390250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191390050 Supplier Thermo Scientific Chemicals Supplier No. 191390050
Add to Cart

Chemical Identifiers

CAS 1229438-47-4
Molecular Formula C21H27Cl4FeN2O
Molecular Weight (g/mol) 521.11

Specifications

Melting Point 174.0°C to 176.0°C
Quantity 5 g
Beilstein 18,V,10,182
Merck Index 15,8118
Formula Weight 521.11
Chemical Name or Material Pyronin B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.