Learn More
Medchemexpress LLC HY-13216 5mg Medchemexpress, Pyroxamide CAS:382180-17-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132165MG
Encompass_Preferred
Medchemexpress, HY-13216 5mg Pyroxamide CAS:382180-17-8 Pyroxamide is a potent inhibitor of histone deacetylase 1 (HDAC1) with an ID50 of 100 nM. Pyroxamide can induce apoptosis and cell cycle arrest in leukemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9837
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.