Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole-2-carboxaldehyde, 99%

Catalog No. AAA1283814
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1283814 25 g
AAA1283809 10 g
AAA1283822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1283814 Supplier Thermo Scientific Chemicals Supplier No. A1283814
Add to Cart

Pyrrole-2-carboxaldehyde, 99%, C5H5NO, CAS Number-1003-29-8, 2-pyrrolecarboxaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolaldehyde, 1h-pyrrole-2-carboxaldehyde, alpha-pyrrolaldehyde, 2-formylpyrrole, 2-pyrrolylcarboxaldehyde, pyrrole-2-aldehyde, pyrrole-2-carboxaldehyde, pyrrole-2-carbaldehyde, 100g | CAS: 1003-29-8 | C5H5NO | 95.101 g/mol

Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carboxaldehyde is used as a pharmaceutical intermediate. It undergoes condensation reaction to produce pyrrole-2-carboxaldehyde salicylhydrazone (PCSH). It is used as a key starting material for the preparation of pyrrole -2-carboxylic acid by oxidation reaction. It is also used to prepare 4-Difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY dyes) through condensation with a pyrrole, which is an important probes for biotechnology and other applications.

Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Store in cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-29-8
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00005217
InChI Key ZSKGQVFRTSEPJT-UHFFFAOYSA-N
Synonym pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde
PubChem CID 13854
ChEBI CHEBI:59978
IUPAC Name 1H-pyrrole-2-carbaldehyde
SMILES C1=CNC(=C1)C=O

Specifications

Melting Point 40°C to 46°C
Boiling Point 217°C to 219°C
Flash Point 106°C (222°F)
Quantity 25 g
Beilstein 105745
Sensitivity Air sensitive
Solubility Information Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water.
Formula Weight 95.1
Percent Purity 99%
Chemical Name or Material Pyrrole-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-705-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.