Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole-3-carboxaldehyde, 97%

Catalog No. AAH3555903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3555903 1 g
AAH35559MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3555903 Supplier Thermo Scientific Chemicals Supplier No. H3555903
Add to Cart

CAS: 7126-39-8 | C5H5NO | 95.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7126-39-8
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD03426733
InChI Key CHNYVNOFAWYUEG-UHFFFAOYSA-N
Synonym pyrrole-3-carboxaldehyde, 1h-pyrrole-3-carboxaldehyde, pyrrole-3-carbaldehyde, 3-formylpyrrole, 3-formyl-1h-pyrrole, 3-formyl-pyrrole, pyrrol-3-aldehyde, pyrrole-3-aldehyde, pubchem18051, acmc-209oht
PubChem CID 640308
IUPAC Name 1H-pyrrole-3-carbaldehyde
SMILES O=CC1=CNC=C1

Specifications

Melting Point 65°C to 68°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 95.1
Percent Purity 97%
Chemical Name or Material Pyrrole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.