Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PZM21
GREENER_CHOICESDP

Catalog No. 721850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-185-0 50 mg
72-181-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 72-185-0 Supplier Tocris Bioscience™ Supplier No. 7218/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective μ receptor agonist

PZM21 is a selective μ receptor agonist (EC50 = 4.6 nM). PZM21 has been reported to exhibit bias for the Gi signaling pathway over arrestin-3. The compound exhibits 500-fold selectivity for μ over δ receptors and no detectable activity at κ or nociceptin receptors and a panel of 316 other GPCRs. PZM21 induces analgesia in a mouse hot plate assay, but not a tail-flick assay and exhibits no significant conditioned place preference response in in vivo assays. CNS penetrant.

Specifications

Chemical Name or Material N-[(2 S)-2-(Dimethylamino)-3-(4-hydroxyphenyl)propyl]-N'-[(1 S)-1-methyl-2-(3-thienyl)ethyl]urea
CAS 1997387-43-5
Quantity 50 mg
Target mu Opioid Receptor Agonists
Molecular Formula C19H27N3O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.