Learn More
OraSure Technologies Q.E.D.™ Saliva Alcohol Test, A-150 Ethanol Control
SDP
Catalog No. 23222111 Shop All OraSure Technologies Products
Change view
Click to view available options
Quantity:
2 x 5 mL
Catalog No. Quantity
23-222-111 2 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-222-111 Supplier OraSure Technologies Supplier No. 31150S
This item is not returnable. View return policy
The Q.E.D.™ A150 Ethanol Controls are intended to be used as a performance control in the determination of alcohol in saliva.
  • Reliable, Fast, Simple
  • Non-invasive
  • Quantitative results in minutes
  • Reads like a thermometer
  • No instrumentation
  • Long shelf life
  • Built-in QA Spot™ assures quality control for every test
Specifications
Certifications/Compliance CLIA-waived and DOT-approved alcohol testing
Type Saliva Alcohol Test Control
For Use With (Application) On-site, low-cost alternative to breath or blood alcohol testing
Quantity 2 x 5 mL