Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Q-Sep QuEChERS dSPE Tubes for Extraction Cleanup
SDP

Catalog No. 06611914 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
06-611-914 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-914 Supplier Restek™ Supplier No. 26221
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fast, simple sample prep for multiresidue pesticide analysis

  • Packaged in foil subpacks of ten for enhanced protection and storage stability
  • Ready to use tubes
    • No glassware required
  • Pre-weighed, ultra-pure sorbents
  • Support original unbuffered, AOAC (2007.01), European (EN 15662) and mini-multiresidue QuEChERS methods

Specifications

For Use With (Application) Extract Cleanup
Type Q-Sep QuEChERS dSPE Tubes
Volume (Metric) 15 mL
Material 1200 mg MgSO4, 400 mg PSA, 400 mg C18-EC
Method AOAC 2007.01
Quantity 50 Pk., Subpacks of 10
Sample Type Foodstuffs with fats and waxes (e.g., cereals, avocado, nuts, seeds and dairy)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.