Learn More
Restek™ Q-Sep QuEChERS Extraction Salts Kit
SDP
Catalog No. 06611961 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
06-611-961 50 Pk.
1 options
Catalog No. 06-611-961 Supplier Restek™ Supplier No. 25848
Extraction salts, unbuffered original method
  • Free-flowing salts transfer easily and completely
  • Easy-open packets eliminate need for second empty tube for salt transfer
  • Convenient slim packets fit perfectly into tubes to prevent spills
  • Ready-to-use tubes
    • No glassware required
  • Pre-weighed, ultra-pure extraction salts
  • Ideal for original unbuffered, AOAC (2007.01) and European (EN 15662) QuEChERS methods
Specifications
Type Q-Sep QuEChERS Extraction Kit
Includes 50 packets and 50 tubes
Material 4 g MsSO4, 1 g NaCl with 50 mL centrifuge tube
Method Original unbuffered
Quantity 50 Pk.