Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Q-VD-OPH, MP Biomedicals™

Catalog No. p-7083071 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3OPH10901 1 mg
MP3OPH10903 3 mg
MP3OPH10905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP3OPH10901 Supplier MP Biomedicals™ Supplier No. 03OPH10901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifically designed for in-vivo research. Q-VD-OPH, MP Biomedicals is synthesized using proprietary technology as a cell-permeable, irreversible caspase inhibitor. The OPH trap, with its superior potency, proven cell permeability and minimal toxicity, p

The Q-VD-OPH irreversibily binds to activated Caspases to block apoptosis. Apoptosis is a physiological form of cell death, which plays an important role in embryogenesis, cellular homoestasis, tissue atrophy, and removal of damaged and mutated cells.

Caspases, which act as molecular chainsaw, are the central components of apoptosis response. Caspases are Cysteine proteases that cleave after aspartate residue in their substrates. At least 7 of 14 known mammalian caspases have important role in apoptosis.

  • Exceptional alternative to the Fluoromethylketone family of inhibitors (these compounds form the basis for important new drug candidates for a wide variety of disease targets)
  • Nontoxic&mdash:preliminary study showed no evidence of toxicity in mice injected intraperitoneally with more than 1000mg/kg
  • Improved potency—IC50 values against purified caspases are in the nanomolar range; in an in vitro cell based assay, greater than 90% inhibition of apoptosis was achieved)
  • Optimal dilutions should be determined by the laboratory for each specific application
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 513.49
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.