Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qa-2 Monoclonal Antibody (69H1-9-9), FITC, eBioscience™
GREENER_CHOICE

Catalog No. 509960
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-996-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-996-0 Supplier Invitrogen™ Supplier No. 11599682
  • $187.00
    $140.65 / Each of 1

    Save $46.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The 69H1-9-9 monoclonal antibody reacts with the mouse Qa-2 molecule, a GPI-linked MHC class Ibeta surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level is different depending on the laboratory mouse strain. Applications Reported: The 69H1-9-9 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 69H1-9-9 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Qa-2 is a GPI-linked MHC class I beta surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level varies between laboratory mouse strains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Qa-2
Applications Flow Cytometry
Classification Monoclonal
Clone 69H1-9-9
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Qa2
Gene Accession No. 0
Gene Alias H2Q7; H2-Qa2; Ped; Qa lymphocyte antigen 2 region; Qa2; Qa-2; Qa-2 antigen
Gene Symbols Qa2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111458
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.