Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qa-2 Mouse, anti-Mouse, R718, Clone: 1-1-2, BD OptiBuild™

Catalog No. BDB751930 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751930 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB751930 Supplier BD Biosciences Supplier No. 751930
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 1-1-2 monoclonal antibody specifically recognizes the Qa-2 MHC class Ib antigen, which is expressed at higher levels on mature T lymphocytes than on B cells. Its expression is upregulated in the late stages of T-cell maturation. Qa-2 is a 40 kDa GPI-linked cell surface molecule encoded by the Q5, Q6, Q7, and/or Q9 genes of the H-2 complex. A 39 kDa soluble form results from alternative splicing of the transcript. Although Qa-2 is nonpolymorphic, its level of expression on both T and B lymphocytes varies among inbred mouse strains. High expressors include strains carrying the H-2[b] haplotype low expressors include H-2[d] strains, and H-2[k] strains are Qa-2-negative.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Qa-2
Applications Flow Cytometry
Classification Monoclonal
Clone 1-1-2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.