Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ QA/QC Microorganism Preparations in KWIK-STIK™ Plus
SDP

Catalog No. 230488X Shop All Microbiologics Products
Change view
Click to view available options
Species:
Aspergillus brasiliensis
Bacillus subtilis
Burkholderia cepacia
Candida albicans
Enterobacter aerogenes
Enterococcus faecalis
Escherichia coli
Pseudomonas aeruginosa
Pseudomonas protegens
Salmonella typhimurium
Staphylococcus aureus
Staphylococcus epidermidis
Streptococcus pyogenes
ATCC®:
10231
12228
13048
14028
16404
19615
25416
25922
25923
27853
29212
6538
6633
8739
9027
ATCC 17386
Biosafety Level:
1
2
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species ATCC® Biosafety Level
23-0488X Burkholderia cepacia 25416 2
23-0385X Streptococcus pyogenes 19615 2
23-0363X Salmonella typhimurium 14028 2
23-0306X Enterobacter aerogenes 13048 2
23-0486X Bacillus subtilis 6633 2
23-0353X Pseudomonas aeruginosa 27853 2
23-017-207 Pseudomonas protegens ATCC 17386 1
23-0335X Escherichia coli 25922 2
23-0371X Staphylococcus epidermidis 12228 2
23-0483X Escherichia coli 8739 2
23-0485X Staphylococcus aureus 6538 2
23-0392X Aspergillus brasiliensis 16404 1
23-0443X Candida albicans 10231 2
23-0366X Enterococcus faecalis 29212 2
23-0360X Staphylococcus aureus 25923 2
23-0484X Pseudomonas aeruginosa 9027 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 23-0488X Supplier Microbiologics™ Supplier No. 0488X
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Come in canisters of five units of a single strain

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

‡

Biotype G

Specifications

ATCC® 25416
Biosafety Level 2
Format KWIK-STIK™ Plus
Product Type KWIK-STIK
Packaging Type Canister
Quantity 5 units of a single strain
Species Burkholderia cepacia
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.