Learn More
Teknova 20x PBS with 0.05% Tween-20. 500mL, Sterile. 8 pack.
Supplier: Teknova P11918PK
2.7M Sodium Chloride, 54mM Potassium Chloride, 28mM Potassium Phosphate Monobasic Anhydrous, 86mM Sodium Phosphate Dibasic Anhydrous, 0.05% Tween 20
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.