Specifications
| Quantity | 400 Tests |
| Includes | One 120mL bottle of F.A.S.T. Auramine O stain and one 120mL bottle of F.A.S.T. decolorizer/quencher |
| For Use With (Application) | Detection of tuberculosis acid fast Bacilli |
| Stain Type | Auramine O |
| Product Type | Auramine Stain |