Learn More
QBC Diagnostics F.A.S.T.™ Consumables
Catalog No. 23900147 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Quantity:
12800 Tests
400 Pack
400 Tests
418 Pack
432 Pack
5 Boxes
675mL
800 Tests
Product Type:
Applicator Stick
Auramine Stain
Microscope Slide
Quality Control Slide
Sputum Cup
Sputum Digestant
Sputum Preparation Tube
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23-900-147 800 Tests Auramine Stain
23-900-151 400 Pack Sputum Cup
23-900-150 432 Pack Microscope Slide
23-900-154 418 Pack Sputum Preparation Tube
23-900-152 400 Pack Applicator Stick
23-900-146 400 Tests Auramine Stain
23-900-148 12800 Tests Auramine Stain
23-900-149 5 Boxes Quality Control Slide
23-900-153 675mL Sputum Digestant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 23-900-147 Supplier QBC Diagnostics Supplier No. 427424
For detection of Tuberculosis Acid Fast Bacilli (AFB)
Specifications
Quantity 800 Tests
Includes One 250mL bottle of F.A.S.T. Auramine O stain and one 250mL bottle of F.A.S.T. decolorizer/quencher
For Use With (Application) Detection of tuberculosis acid fast Bacilli
Stain Type Auramine O
Product Type Auramine Stain