Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QBC Diagnostics F.A.S.T.™ Consumables

Catalog No. 23900150 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Quantity:
12800 Tests
400 Pack
400 Tests
418 Pack
432 Pack
5 Boxes
675mL
800 Tests
Product Type:
Applicator Stick
Auramine Stain
Microscope Slide
Quality Control Slide
Sputum Cup
Sputum Digestant
Sputum Preparation Tube
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23-900-150 432 Pack Microscope Slide
23-900-151 400 Pack Sputum Cup
23-900-154 418 Pack Sputum Preparation Tube
23-900-152 400 Pack Applicator Stick
23-900-146 400 Tests Auramine Stain
23-900-147 800 Tests Auramine Stain
23-900-148 12800 Tests Auramine Stain
23-900-149 5 Boxes Quality Control Slide
23-900-153 675mL Sputum Digestant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 23-900-150 Supplier QBC Diagnostics Supplier No. 427411

For detection of Tuberculosis Acid Fast Bacilli (AFB)

Specifications

Quantity 432 Pack
Includes 1 box of 432 slides
Format Slide
For Use With (Application) Finding the proper focal plane and maintaining focus throughout review
Product Type Microscope Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.