Learn More
Specifications
Specifications
| Quantity | 400 Pack |
| Includes | 400 applicator sticks |
| Format | Stick |
| For Use With (Application) | Transferring sputum smears to the F.A.S.T. SureFocus Slide during the staining process |
| Product Type | Applicator Stick |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.