Learn More
QBC Diagnostics ParaLens Advance™ Fluorescence Microscope System: With 60X, 100X Obj. and With Portability Pack
Catalog No. 23900113 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Magnification Power:
100X
20X, 40X, 60X, 100X
Includes:
Main body assembly with 100X objective and one 30mL bottle of immersion oil
Portability pack, 4 main body assemblies with 20X, 40X, 60X and 100X objectives, one 30mL bottle of immersion oil, ParaViewer™ tube holder, solar cell battery pack, solar cell wall plug, USB to light source cable, 12V battery clip to lig
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power Includes
23-900-113 20X, 40X, 60X, 100X Portability pack, 4 main body assemblies with 20X, 40X, 60X and 100X objectives, one 30mL bottle of immersion oil, ParaViewer™ tube holder, solar cell battery pack, solar cell wall plug, USB to light source cable, 12V battery clip to lig
23-900-116 100X Main body assembly with 100X objective and one 30mL bottle of immersion oil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-900-113 Supplier QBC Diagnostics Supplier No. 424333
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Affordable fluorescence microscopy on any microscope
The QBC ParaLens Advance is a microscope attachment that utilizes a durable, long-lasting LED light source to provide fluorescence capabilities to any light microscope. The light source can be powered by an AC to DC power pack, a solar powered battery pack, a USB port or other convenient power source.
ParaLens Advance with 60X Objective
  • Designed for malaria detection using the QBC Malaria Test
  • Includes main body assembly with 60X objective, blue LED light source, detachable filter set arm for use with blue LED light source, power pack with international plug adapters, one 30mL bottle of immersion oil, ParaViewer™, flat blade screwdriver, and ParaLens Advance preventative maintenance kit
  • Packaged in a hard shell carrying case
ParaLens Advance with 100X Objective
  • Includes main body assembly with 100X objective, blue LED light source, detachable filter set arm for use with blue LED light source, power pack with international plug adapters, one 30mL bottle of immersion oil, ParaViewer, flat blade screwdriver, and ParaLens Advance preventative maintenance kit
  • Packaged in a hard shell carrying case
ParaLens Advance Complete Fluorescence System with Portability Pack
  • Includes four main body assemblies with 20X, 40X, 60X and 100X objectives, blue LED light source, detachable filter set arm for use with blue LED light source, power pack with international plug adapters, one 30mL bottle of immersion oil, ParaViewer, flat blade screwdriver, ParaLens Advance preventative maintenance kit, solar cell battery pack, solar cell wall plug, USB to 3.5mm cable, 3.5mm to light source cable, 12V battery clip to lighter adapter and lighter adapter to light source cable
  • Packaged in a hard shell carrying case
Specifications
Includes Portability pack, 4 main body assemblies with 20X, 40X, 60X and 100X objectives, one 30mL bottle of immersion oil, ParaViewer™ tube holder, solar cell battery pack, solar cell wall plug, USB to light source cable, 12V battery clip to lig
Magnification Power 20X, 40X, 60X, 100X
For Use With (Application) Malaria detection using the QBC malaria test
Port Type USB
Lighting Type LED, Blue