Learn More
Beckman Coulter Life Sciences QbD1200 + TOC Analyzer
Catalog No. 06713247
Click to view available options
Type:
Elemental Analyzer
Catalog No. Type
06-713-247 Elemental Analyzer
1 options
Catalog No. 06-713-247 Supplier Beckman Coulter Life Sciences Supplier No. 9880100
Add to Cart
Add to Cart
Request a Quote
The QbD1200+ TOC Analyzer is designed to make testing simple, reproducible and intuitive.
  • QbD1200+ Laboratory TOC Analyzer with integrated 10.4 in touchscreen.
  • Digital NDIR technology with auto-correct drift ensuring long-term stability.
  • Dynamic endpoint detection technology ensuring removal of the inorganic carbon and full oxidation of the organic matter.
  • Automated calibration, system suitability testing and report generation.
  • Self health checks on instrument startup and continuous monitoring of instrument health status.
  • Designed to support 21 CFR Part 11 compliant workflows and global Pharmacopeia regulations, including USP, JP, EP, IP, KP, and ICH
  • 100-240 VAC, 50-60 Hz, US power supply
  • Includes Analyzer, power cord, power supply, empty reagent bottle, gas inlet tube and GLS water vessel.
TRUSTED_SUSTAINABILITY
Specifications
Type Elemental Analyzer
Detector Type NDIR
For Use With (Application) Pharmaceuticals
Voltage 100/240 VAC
Peace of Mind, Simplify Workflows, Low Total Cost of Ownership