Learn More
Cole-Parmer Port Disk Hose Barb Fitting, Natural Kynar Flex, No Dimples, Cleanroom Packed, 1/8" ID; 10/Pk
Supplier: Cole-Parmer 5011716
Use these high-quality PVC-free port disks with hose barb to transfer fluids, buffers, media, and drug compounds from single-use bags used in pharmaceutical and medical industries. Ultrasonic, radio frequency, or thermal weld port disks to Kynar medical and bioreactor bags to easily transfer fluids.
Natural Kynar Flex material exhibits excellent chemical resistance, UV resistance, and high purity. Kynar Flex offers slightly more flexibility compared to regular Kynar, minimizing the risk of it cracking with sideload.
Disks are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.