Learn More
Thermo Scientific™ AcroMetrix NAT Blood Screening Controls
Catalog No. 23279180 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix WNVRNA Control
Detectable Analytes:
West Nile Virus (WNV)
Catalog No. Description Detectable Analytes
23-279-180 AcroMetrix WNVRNA Control West Nile Virus (WNV)
1 options
Catalog No. 23-279-180 Supplier Thermo Scientific™ Supplier No. 960500
Assess performance of NAT assays that test human plasma from donations of whole blood and blood components with these controls, which are for In Vitro Diagnostic Use.
Estimate the precision and reproducibility of NAT assays that are designed to detect viral nucleic acids in human plasma from donations of whole blood and blood components for transfusion with the Thermo Scientific™ AcroMetrix NAT Blood Screening Controls. These controls mimic naturally occurring plasma specimens and can help detect potential systematic deviations of tests for the qualitative determination of viral nucleic acids. The controls are for In Vitro Diagnostic Use.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix WNVRNA Control
Detectable Analytes West Nile Virus (WNV)
Sample Type Plasma
Type Control
Packaging 10 x 1.4mL
Quantity 10 x 1.4 mL