Learn More
QIAGEN QIAcard FTA Mini
Catalog No. 09800994
Change view
Click to view available options
Quantity:
100 Pk.
25 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-800-994 25 Pk.
09-923-342 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-800-994 Supplier QIAGEN Supplier No. WB120355
Add to Cart
Add to Cart
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
  • QIAcard FTA Mini format with 2 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Description Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards.
Sample Type Blood
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
No. of Samples 2
Product Type FTA Mini Card
Quantity 25 Pk.
Sample Volume 125 μL