Learn More
QKI Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300183A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, Chicken, Bovine, Dog, Horse, Pig and Cat
QKI belongs to a family of RNA-binding proteins that have an HNRNPK (MIM 600712) homology (KH) domain embedded in a 200-amino acid region called the GSG domain. Other members of this family include SAM68 (KHDRBS1; MIM 602489) and SF1 (MIM 601516) (Chen and Richard, 1998 [PubMed 9671495]). QKI proteins regulate RNA splicing, export of target RNAs from the nucleus, translation of proteins, and RNA stability (Lauriat et al., 2008 [PubMed 17918747]).Specifications
QKI | |
Polyclonal | |
Unconjugated | |
QKI | |
Protein quaking; HqkI; Hqk; QKI; HKQ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19317, 9444 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96PU8, Q9QYS9 | |
Qk, QKI | |
C-terminus of isoform 5 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.