Learn More
Medchemexpress LLC HY-114388 5mg Medchemexpress, QM385 CAS:2093421-02-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1143885MG
Encompass_Preferred
Medchemexpress, HY-114388 5mg QM385 CAS:2093421-02-2 QM385 is a potent sepiapterin reductase (SPR) inhibitor with an IC50 of 1.49 nM, which blocks T-cell proliferation and autoimmunity at nanomolar potency and with good oral bioavailability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9790
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.